Assay Design
Gene Transcripts
- Gene Symbol
- CRYBB1
- Entrez Gene ID
-
374000
- Gene Name
- crystallin beta B1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 7465326 - 7467621 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204180.1 |
NP_989511.1 |
2 - 3 |
215 |
60 |
GenBank mRNA |
M11619.1 |
- |
2 - 3 |
215 |
60 |
View Full Product Details