Assay Design
Gene Transcripts
- Gene Symbol
- SMAD6
- Entrez Gene ID
-
374096
- Gene Name
- SMAD family member 6
- Gene Aliases
-
MADH6
- Chromosome Location
-
Chr.10: 19114126 - 19141544 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF165889.1 |
- |
2 - 3 |
978 |
62 |
FJ417094.1 |
- |
2 - 3 |
798 |
62 |
View Full Product Details