Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_204265.1 | NP_989596.1 | 4 - 5 | 810 | 121 |
XM_015294110.2 | XP_015149596.1 | 5 - 6 | 477 | 121 | |
XM_015294111.2 | XP_015149597.1 | 5 - 6 | 439 | 121 | |
XM_015294113.2 | XP_015149599.1 | 5 - 6 | 379 | 121 | |
XM_015294114.2 | XP_015149600.1 | 5 - 6 | 444 | 121 | |
GenBank mRNA | AB069964.1 | - | 4 - 5 | 810 | 121 |
AF461016.1 | - | 5 - 6 | 372 | 121 |