Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_204288.1 | NP_989619.1 | 2 - 3 | 171 | 53 |
XM_015292934.2 | XP_015148420.1 | 2 - 3 | 154 | 53 | |
XM_015292935.1 | XP_015148421.1 | 2 - 3 | 154 | 53 | |
XM_025154402.1 | XP_025010170.1 | 2 - 3 | 154 | 53 | |
XM_025154403.1 | XP_025010171.1 | 2 - 3 | 207 | 53 | |
GenBank mRNA | AF439559.1 | - | 2 - 3 | 171 | 53 |