Assay Design



Gene Transcripts

Gene Symbol
PAFAH1B1
Entrez Gene ID
374224
Gene Name
platelet activating factor acetylhydrolase 1b regulatory subunit 1
Gene Aliases
-
Chromosome Location
Chr.19: 9585935 - 9615086 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204324.1 NP_989655.1 8 - 9 1128 68
XM_015295659.2 XP_015151145.1 8 - 9 1109 68
XM_015295660.1 XP_015151146.1 9 - 10 1129 68
XM_015295661.2 XP_015151147.1 9 - 10 1297 68
XM_025141880.1 XP_024997648.1 10 - 11 8008 68
XM_025141881.1 XP_024997649.1 10 - 11 8033 68
GenBank mRNA AF113946.1 - 8 - 9 1128 68

View Full Product Details