Assay Design



Gene Transcripts

Gene Symbol
BMP2
Entrez Gene ID
378779
Gene Name
bone morphogenetic protein 2
Gene Aliases
-
Chromosome Location
Chr.3: 15201425 - 15437861 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204358.1 NP_989689.1 1 - 2 331 67
XM_015283435.2 XP_015138921.1 6 - 7 2694 67
XM_025148482.1 XP_025004250.1 9 - 10 2698 67
XM_025148483.1 XP_025004251.1 8 - 9 2641 67
XM_025148484.1 XP_025004252.1 9 - 10 2695 67
XM_025148485.1 XP_025004253.1 10 - 11 2788 67
XM_025148486.1 XP_025004254.1 7 - 8 1438 67
XM_025148487.1 XP_025004255.1 8 - 9 1528 67
XM_025148488.1 XP_025004256.1 8 - 9 2638 67
GenBank mRNA AY237249.1 - 1 - 2 331 67

View Full Product Details