Assay Design



Gene Transcripts

Gene Symbol
HMGCR
Entrez Gene ID
395145
Gene Name
3-hydroxy-3-methylglutaryl-CoA reductase
Gene Aliases
HMGR
Chromosome Location
Chr.Z: 24007046 - 24025405 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204485.2 NP_989816.2 2 - 3 309 71
XM_015277227.2 XP_015132713.1 2 - 3 221 71
GenBank mRNA AB109635.1 - 1 - 2 154 71
AJ720311.1 - 2 - 3 284 71

View Full Product Details