Assay Design



Gene Transcripts

Gene Symbol
RAB5C
Entrez Gene ID
395197
Gene Name
RAB5C, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.27: 7656878 - 7667208 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204525.1 NP_989856.1 3 - 4 450 114
XM_015299568.2 XP_015155054.1 4 - 5 589 114
GenBank mRNA Y07811.1 - - 450 114

View Full Product Details