Assay Design
Gene Transcripts
- Gene Symbol
- ITM2B
- Entrez Gene ID
-
395969
- Gene Name
- integral membrane protein 2B
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 170036141 - 170062172 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_205085.2 |
NP_990416.2 |
2 - 3 |
385 |
103 |
GenBank mRNA |
AF017985.1 |
- |
2 - 3 |
247 |
103 |
View Full Product Details