Assay Design



Gene Transcripts

Gene Symbol
PSMD9
Entrez Gene ID
416853
Gene Name
proteasome 26S subunit, non-ATPase 9
Gene Aliases
-
Chromosome Location
Chr.15: 5989102 - 5991824 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006189.1 NP_001006189.1 1 - 2 160 115
GenBank mRNA AJ720363.1 - 1 - 2 160 115
BX935300.1 - 1 - 2 117 115
BX936274.1 - 1 - 2 117 115

View Full Product Details