Assay Design



Gene Transcripts

Gene Symbol
NELFB
Entrez Gene ID
417254
Gene Name
negative elongation factor complex member B
Gene Aliases
COBRA1
Chromosome Location
Chr.17: 2266800 - 2277936 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006202.1 NP_001006202.1 3 - 4 404 102
GenBank mRNA AJ719725.1 - 3 - 4 404 102

View Full Product Details