Assay Design



Gene Transcripts

Gene Symbol
PCNP
Entrez Gene ID
418398
Gene Name
PEST proteolytic signal containing nuclear protein
Gene Aliases
-
Chromosome Location
Chr.1: 86433447 - 86447140 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006253.1 NP_001006253.1 1 - 2 80 127
GenBank mRNA AJ719633.1 - 1 - 2 80 127

View Full Product Details