Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001006323.1 | NP_001006323.1 | - | 428 | 68 |
XM_015298930.2 | XP_015154416.1 | - | 424 | 68 | |
XM_025143692.1 | XP_024999460.1 | - | 1952 | 68 | |
GenBank mRNA | AJ719588.1 | - | - | 428 | 68 |
AJ721138.1 | - | - | 394 | 68 |