Assay Design



Gene Transcripts

Gene Symbol
XRCC3
Entrez Gene ID
423485
Gene Name
X-ray repair cross complementing 3
Gene Aliases
-
Chromosome Location
Chr.5: 50666213 - 50674947 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006489.1 NP_001006489.1 6 - 7 1000 69
XM_015287481.2 XP_015142967.1 6 - 7 1004 69
XM_025150795.1 XP_025006563.1 5 - 6 850 69
GenBank mRNA AJ719558.1 - 6 - 7 1000 69
BX935798.2 - 6 - 7 1001 69

View Full Product Details