Assay Design



Gene Transcripts

Gene Symbol
STX10
Entrez Gene ID
424413
Gene Name
syntaxin 10
Gene Aliases
STX6, syntaxin-6
Chromosome Location
Chr.8: 6189807 - 6203803 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006531.1 NP_001006531.1 2 - 3 235 72
GenBank mRNA AJ719915.1 - 2 - 3 235 72

View Full Product Details