Assay Design



Gene Transcripts

Gene Symbol
NDUFB5
Entrez Gene ID
424978
Gene Name
NADH:ubiquinone oxidoreductase subunit B5
Gene Aliases
-
Chromosome Location
Chr.9: 17584886 - 17588422 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006553.1 NP_001006553.1 4 - 5 340 87
GenBank mRNA AJ720223.1 - 4 - 5 340 87

View Full Product Details