Assay Design



Gene Transcripts

Gene Symbol
WDR45B
Entrez Gene ID
417339
Gene Name
WD repeat domain 45B
Gene Aliases
WDR45L
Chromosome Location
Chr.18: 3388196 - 3404562 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007844.1 NP_001007845.1 6 - 7 730 86
XM_015295140.2 XP_015150626.1 7 - 8 2920 86
XM_015295142.2 XP_015150628.1 5 - 6 2671 86
GenBank mRNA AJ719918.1 - 6 - 7 730 86

View Full Product Details