Assay Design



Gene Transcripts

Gene Symbol
GCLM
Entrez Gene ID
424492
Gene Name
glutamate-cysteine ligase modifier subunit
Gene Aliases
-
Chromosome Location
Chr.8: 14159785 - 14170017 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007953.1 NP_001007954.1 6 - 7 721 109
GenBank mRNA AJ719931.1 - 6 - 7 721 109

View Full Product Details