Assay Design



Gene Transcripts

Gene Symbol
RASSF5
Entrez Gene ID
419844
Gene Name
Ras association domain family member 5
Gene Aliases
-
Chromosome Location
Chr.26: 2489020 - 2506554 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008452.1 NP_001008452.1 - 401 106
GenBank mRNA AJ720993.1 - - 401 106

View Full Product Details