Assay Design



Gene Transcripts

Gene Symbol
COMMD5
Entrez Gene ID
422376
Gene Name
COMM domain containing 5
Gene Aliases
-
Chromosome Location
Chr.4: 16711115 - 16731238 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008465.1 NP_001008465.1 4 - 5 409 88
GenBank mRNA AJ719495.1 - 4 - 5 409 88

View Full Product Details