Assay Design
Gene Transcripts
- Gene Symbol
- CHRNA3
- Entrez Gene ID
-
386578
- Gene Name
- cholinergic receptor nicotinic alpha 3 subunit
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 4073466 - 4079283 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204416.1 |
NP_989747.1 |
4 - 5 |
356 |
117 |
GenBank mRNA |
M37336.1 |
- |
4 - 5 |
356 |
117 |
View Full Product Details