Assay Design
Gene Transcripts
- Gene Symbol
- CHRNA9
- Entrez Gene ID
-
395522
- Gene Name
- cholinergic receptor nicotinic alpha 9 subunit
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 68964832 - 68969315 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204760.1 |
NP_990091.1 |
2 - 3 |
215 |
115 |
GenBank mRNA |
AF082192.1 |
- |
2 - 3 |
215 |
115 |
View Full Product Details