Assay Design
Gene Transcripts
- Gene Symbol
- LY6E
- Entrez Gene ID
-
395550
- Gene Name
- lymphocyte antigen 6 family member E
- Gene Aliases
-
SCA-2, SCA2
- Chromosome Location
-
Chr.2: 148338604 - 148344697 on Build GRCg6a
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_204775.1 |
NP_990106.1 |
2 - 3 |
228 |
75 |
GenBank mRNA |
L34554.1 |
- |
2 - 3 |
228 |
75 |
View Full Product Details