Assay Design



Gene Transcripts

Gene Symbol
CYP2C18
Entrez Gene ID
414833
Gene Name
cytochrome P450 family 2 subfamily C member 18
Gene Aliases
CYP2C45
Chromosome Location
Chr.6: 17408110 - 17413208 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001752.2 NP_001001752.1 8 - 9 1316 141
GenBank mRNA AJ430583.1 - 8 - 9 1317 141

Gene Symbol
RP11-400G3.5
Entrez Gene ID
100858007
Gene Name
Uncharacterized protein
Gene Aliases
CYP2C21L
Chromosome Location
Chr.6: 17399196 - 17404444 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015288526.2 XP_015144012.1 8 - 9 1330 141

View Full Product Details