Assay Design



Gene Transcripts

Gene Symbol
EXOC8
Entrez Gene ID
421531
Gene Name
exocyst complex component 8
Gene Aliases
-
Chromosome Location
Chr.3: 39848318 - 39851256 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006404.1 NP_001006404.1 1 - 2 1560 120
GenBank mRNA AJ720591.1 - 1 - 2 1560 120

View Full Product Details