Assay Design



Gene Transcripts

Gene Symbol
OR6B1
Entrez Gene ID
415340
Gene Name
olfactory receptor family 6 subfamily B member 1
Gene Aliases
COR7A
Chromosome Location
Chr.10: 1207419 - 1208391 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030392.1 NP_001025563.1 1 - 1 253 137

View Full Product Details