Assay Design



Gene Transcripts

Gene Symbol
PNRC1
Entrez Gene ID
421817
Gene Name
proline rich nuclear receptor coactivator 1
Gene Aliases
-
Chromosome Location
Chr.3: 75829794 - 75832049 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012291.1 NP_001012291.1 2 - 2 852 109
GenBank mRNA AJ720799.1 - 2 - 2 852 109

View Full Product Details