Assay Design



Gene Transcripts

Gene Symbol
OPN5
Entrez Gene ID
428670
Gene Name
opsin 5
Gene Aliases
GPR136, GRP136, Opn5m, PGR12, TMEM13, cOpn5m, neuropsin, opsin-5
Chromosome Location
Chr.3: 110092843 - 110119982 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130743.1 NP_001124215.1 3 - 4 341 84
XM_015285151.2 XP_015140637.1 2 - 3 711 84
GenBank mRNA AB368182.1 - 3 - 4 341 84

View Full Product Details