Assay Design



Gene Transcripts

Gene Symbol
OPN5
Entrez Gene ID
428670
Gene Name
opsin 5
Gene Aliases
GPR136, GRP136, Opn5m, PGR12, TMEM13, cOpn5m, neuropsin, opsin-5
Chromosome Location
Chr.3: 110092843 - 110119982 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130743.1 NP_001124215.1 6 - 7 1088 76
XM_015285151.2 XP_015140637.1 5 - 6 1458 76
GenBank mRNA AB368182.1 - 6 - 7 1088 76

View Full Product Details