Assay Design



Gene Transcripts

Gene Symbol
CD14
Entrez Gene ID
100194427
Gene Name
CD14 molecule
Gene Aliases
-
Chromosome Location
Chr.13: 1611909 - 1615529 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001139478.1 NP_001132950.1 1 - 1 751 102
XM_015293508.2 XP_015148994.1 2 - 2 1061 102
GenBank mRNA AM933591.1 - 1 - 1 751 102

View Full Product Details