Assay Design



Gene Transcripts

Gene Symbol
CLPS
Entrez Gene ID
771102
Gene Name
colipase
Gene Aliases
-
Chromosome Location
Chr.26: 101626 - 102258 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145491.1 NP_001138963.1 - 209 103
GenBank mRNA EU407547.1 - - 209 103

View Full Product Details