Assay Design



Gene Transcripts

Gene Symbol
NANOG
Entrez Gene ID
100272166
Gene Name
Nanog homeobox
Gene Aliases
-
Chromosome Location
Chr.1: 75593243 - 75596024 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146142.1 NP_001139614.1 3 - 4 518 70
GenBank mRNA DQ867025.1 - 3 - 4 518 70

View Full Product Details