Assay Design



Gene Transcripts

Gene Symbol
CKM
Entrez Gene ID
396507
Gene Name
creatine kinase, M-type
Gene Aliases
CPK-M
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205507.1 NP_990838.1 - 1149 104
GenBank mRNA M10012.1 - - 1142 104
X00954.1 - - 1149 104

View Full Product Details