Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001250612.1 | NP_001237541.1 | 4 - 5 | 580 | 105 |
GenBank mRNA | AF034210.1 | - | 4 - 5 | 580 | 105 |
AK243764.1 | - | 4 - 5 | 340 | 105 | |
AK244943.1 | - | 4 - 5 | 349 | 105 |