Assay Design
Gene Transcripts
- Gene Symbol
- AO
- Entrez Gene ID
-
547647
- Gene Name
- ascorbate oxidase
- Gene Aliases
-
GmAO
- Chromosome Location
-
Chr.20: 17148271 - 17152533 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF529300.1 |
- |
2 - 3 |
331 |
97 |
View Full Product Details