Assay Design



Gene Transcripts

Gene Symbol
CA1
Entrez Gene ID
547671
Gene Name
carbonic anhydrase
Gene Aliases
-
Chromosome Location
Chr.6: 15634897 - 15638994 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249275.1 NP_001236204.1 6 - 7 537 72
GenBank mRNA AJ239132.1 - 6 - 7 537 72
BT097689.1 - 6 - 7 555 72

View Full Product Details