Assay Design



Gene Transcripts

Gene Symbol
URE
Entrez Gene ID
547672
Gene Name
urease
Gene Aliases
-
Chromosome Location
Chr.11: 38418308 - 38425109 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249285.1 NP_001236214.1 12 - 13 1626 101
GenBank mRNA AJ276866.1 - 13 - 14 1800 101
AY230156.1 - 12 - 13 1626 101

View Full Product Details