Assay Design
Gene Transcripts
- Gene Symbol
- SBP
- Entrez Gene ID
-
606297
- Gene Name
- selenium binding protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.8: 8307249 - 8310573 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ401230.1 |
- |
5 - 6 |
885 |
101 |
View Full Product Details