Assay Design
Gene Transcripts
- Gene Symbol
- SIR
- Entrez Gene ID
-
547693
- Gene Name
- ferredoxin:sulfite reductase precursor
- Gene Aliases
-
DCP68, GmSiR
- Chromosome Location
-
Chr.11: 7051681 - 7056161 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY017473.1 |
- |
2 - 3 |
1034 |
81 |
View Full Product Details