Assay Design



Gene Transcripts

Gene Symbol
AMS1
Entrez Gene ID
547702
Gene Name
beta-amyrin synthase
Gene Aliases
-
Chromosome Location
Chr.7: 128115 - 136350 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249662.1 NP_001236591.1 7 - 8 1094 93
GenBank mRNA AK245291.1 - 7 - 8 1156 93
AY095999.1 - 7 - 8 1094 93

View Full Product Details