Assay Design



Gene Transcripts

Gene Symbol
AMS1
Entrez Gene ID
547702
Gene Name
beta-amyrin synthase
Gene Aliases
-
Chromosome Location
Chr.7: 128115 - 136350 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249662.1 NP_001236591.1 9 - 10 1328 103
GenBank mRNA AK245291.1 - 9 - 10 1390 103
AY095999.1 - 9 - 10 1328 103

View Full Product Details