Assay Design
Gene Transcripts
- Gene Symbol
- GR
- Entrez Gene ID
-
547793
- Gene Name
- glutathione reductase
- Gene Aliases
-
-
- Chromosome Location
-
Chr.2: 14441673 - 14447178 on Build Glycine_max_v2.1
- Gene Symbol
- LOC100810005
- Entrez Gene ID
-
100810005
- Gene Name
- glutathione reductase, chloroplastic-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 2799231 - 2804706 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK286768.1 |
- |
7 - 8 |
1323 |
92 |
View Full Product Details