Assay Design



Gene Transcripts

Gene Symbol
SNAP-1
Entrez Gene ID
547812
Gene Name
nucleosome assembly protein 1
Gene Aliases
-
Chromosome Location
Chr.15: 15484078 - 15487474 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251385.1 NP_001238314.1 2 - 3 168 54
GenBank mRNA BT096713.1 - 2 - 3 201 54
BT098302.1 - 2 - 3 199 54
L38856.1 - 2 - 3 168 54

View Full Product Details