Assay Design
Gene Transcripts
- Gene Symbol
- RAB2
- Entrez Gene ID
-
732635
- Gene Name
- guanine nucleotide regulatory protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.9: 1277986 - 1281220 on Build Glycine_max_v2.1
- Gene Symbol
- LOC100784816
- Entrez Gene ID
-
100784816
- Gene Name
- ras-related protein RABB1c-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.15: 9554351 - 9557776 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK246005.1 |
- |
5 - 6 |
716 |
116 |
View Full Product Details