Assay Design



Gene Transcripts

Gene Symbol
SFR2
Entrez Gene ID
732645
Gene Name
beta-glycosidase-like
Gene Aliases
-
Chromosome Location
Chr.17: 2710451 - 2715424 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251399.1 NP_001238328.1 6 - 7 1197 86
GenBank mRNA AM238658.1 - 6 - 7 1197 86

View Full Product Details