Assay Design



Gene Transcripts

Gene Symbol
MT2
Entrez Gene ID
732644
Gene Name
malonyltransferase
Gene Aliases
-
Chromosome Location
Chr.18: 5067269 - 5071081 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001251383.1 NP_001238312.1 9 - 10 1068 78
GenBank mRNA AK244797.1 - 9 - 10 1113 78
DQ278438.1 - 9 - 10 1068 78

View Full Product Details