Assay Design



Gene Transcripts

Gene Symbol
EN1
Entrez Gene ID
100101913
Gene Name
endonuclease
Gene Aliases
-
Chromosome Location
Chr.15: 5202032 - 5204030 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001248396.1 NP_001235325.1 6 - 7 661 120
GenBank mRNA AK246074.1 - 6 - 7 702 120
EF521872.1 - 6 - 7 661 120

View Full Product Details