Assay Design



Gene Transcripts

Gene Symbol
WNK6
Entrez Gene ID
100272197
Gene Name
with no lysine kinase 6
Gene Aliases
-
Chromosome Location
Chr.7: 4631644 - 4635415 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249016.1 NP_001235945.1 3 - 4 258 90
GenBank mRNA FJ795036.1 - 3 - 4 258 90

View Full Product Details