Assay Design



Gene Transcripts

Gene Symbol
WNK9
Entrez Gene ID
100272199
Gene Name
with no lysine kinase 9
Gene Aliases
-
Chromosome Location
Chr.19: 48932330 - 48935317 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249042.1 NP_001235971.1 3 - 4 331 100
GenBank mRNA FJ795039.1 - 3 - 4 331 100

View Full Product Details