Assay Design
Gene Transcripts
- Gene Symbol
- WRKY8
- Entrez Gene ID
-
100127368
- Gene Name
- WRKY8 protein
- Gene Aliases
-
-
- Chromosome Location
-
Chr.19: 46863027 - 46865461 on Build Glycine_max_v2.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
EU019556.1 |
- |
3 - 3 |
549 |
83 |
View Full Product Details